missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFAF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570777
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 93%; Zebrafish: 92%.
The NADH-ubiquinone oxidoreductase complex (complex I) of the mitochondrial respiratory chain catalyzes the transfer of electrons from NADH to ubiquinone, and consists of at least 43 subunits. The complex is located in the inner mitochondrial membrane. This gene encodes a mitochondrial protein that is associated with the matrix face of the mitochondrial inner membrane and is required for complex I assembly. A mutation in this gene results in mitochondrial complex I deficiency. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| NDUFAF5 | |
| Polyclonal | |
| Unconjugated | |
| NDUFAF5 | |
| 2310003L22Rik; arginine-hydroxylase NDUFAF5, mitochondrial; bA526K24.2; C13H20orf7; C20orf7; C24H20orf7; dJ842G6.1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5; arginine-hydroxylase NDUFAF5, mitochondrial; NADH:ubiquinone oxidoreductase complex assembly factor 5; ndufaf5; probable methyltransferase C20orf7 homolog, mitochondrial; probable methyltransferase C20orf7, mitochondrial; putative methyltransferase C20orf7, mitochondrial-like protein; putative methyltransferase NDUFAF5; RGD1309829; zgc:162919 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 69487 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A2APY7 | |
| NDUFAF5 | |
| synthetic peptide directed towards the middle region of mouse NDUFAF5. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction