missing translation for 'onlineSavingsMsg'
Learn More

Nebivolol, TRC

Product Code. 17984405
Change view
Click to view available options
Quantity:
10 mg
250 mg
Unit Size:
10mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984405 10 mg 10mg
30136897 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17984405 Supplier Toronto Research Chemicals Supplier No. TRCN38793310MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99200-09-6
Molecular Formula C22H25F2NO4
Molecular Weight (g/mol) 405.44
Synonym α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol];
IUPAC Name 1-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)-2-[[2-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)-2-hydroxyethyl]amino]ethanol
SMILES FC1=CC=C2C(CCC(C(O)CNCC(O)C3CCC4=CC(F)=CC=C4O3)O2)=C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H25F2NO4/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2
Formula Weight 405.1752
Percent Purity >95%
Chemical Name or Material Nebivolol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.