Learn More
NEDD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13993-1-AP-20UL
 View more versions of this product
Product Code. 16852454
  • 145.00€ / 20µL
Description
This antibody works well in WB, IHC, IP and IF application.
Neural precursor cell expressed developmentally down-regulated protein 1 (NEDD1) is required for mitosis progression. NEDD1 promotes the nucleation of microtubules from the spindle.
Specifications
Specifications
NEDD1
Polyclonal
Unconjugated
NEDD1
GCP WD, NEDD 1, NEDD1, Protein NEDD1, TUBGCP7
Rabbit
Antigen Affinity Chromatography
RUO
121441, 17997, 299730
-20°C
Liquid
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P33215, Q8NHV4
NEDD1
NEDD1 Fusion Protein Ag5077
20 μL
Primary
Human, Rat, Mouse