missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEDD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68758-25ul
This item is not returnable.
View return policy
Description
NEDD8 Polyclonal antibody specifically detects NEDD8 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| NEDD8 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| FLJ43224, MGC104393, MGC125896, MGC125897, NEDD-8, Neddylin, Neural precursor cell expressed developmentally down-regulated protein 8, neural precursor cell expressed, developmentally down-regulated 8, Ubiquitin-like protein Nedd8 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MLIKVKTLTGKEIEIDIEPTDKVERIKERVEEKEGI | |
| 25 μL | |
| Cancer | |
| 4738 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction