Learn More
Needle Cleaning Solution
Product Code. 11930557
Change view
Click to view available options
For Use With (Equipment):
Osmometer Type 15/15M and 16/16M
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11930557 Osmometer Type 15/15M and 16/16M Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11930557 Supplier Loeser Supplier No. 0.01.0004
  • ONLINE EXCLUSIVE PRICE
    7.10€
    4.90€ / Pack of 1
    Save 2.20€
    30% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Needle cleaning solution for i Osmometer
  • Solution for cleaning the needle hole of automatic Osmometers
  • 20mL bottle
Specifications
Product Type Needle Cleaning Solution
For Use With (Equipment) Osmometer Type 15/15M and 16/16M
Volume (Metric) 20 mL