missing translation for 'onlineSavingsMsg'
Learn More

Nefopam Hydrochloride, TRC

Product Code. 17959849
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959849 250 mg 250mg
18071120 10 mg 10mg
18053540 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17959849 Supplier Toronto Research Chemicals Supplier No. TRCN389400250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23327-57-3
Molecular Formula C17 H19 N O . Cl H
Molecular Weight (g/mol) 289.8
Synonym 1H-2,5-Benzoxazocine, 3,4,5,6-tetrahydro-5-methyl-1-phenyl-, hydrochloride (1:1), 1H-2,5-Benzoxazocine, 3,4,5,6-tetrahydro-5-methyl-1-phenyl-, hydrochloride (8CI,9CI), (±)-Nefopam hydrochloride, Acupan, Ajan, Fenazoxine, Fenazoxine hydrochloride, Nefopam hydrochloride, R 738
IUPAC Name 5-methyl-1-phenyl-1,3,4,6-tetrahydro-2,5-benzoxazocine;hydrochloride
SMILES Cl.CN1CCOC(c2ccccc2)c3ccccc3C1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H19NO.ClH/c1-18-11-12-19-17(14-7-3-2-4-8-14)16-10-6-5-9-15(16)13-18;/h2-10,17H,11-13H2,1H3;1H
Formula Weight 289.1233
Percent Purity >95%
Chemical Name or Material Nefopam Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.