Learn More
Invitrogen™ Neisseria gonorrhoeae Monoclonal Antibody (GC12-316.4), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA5-46064
 View more versions of this product
Product Code. 17800406
  • 424.00€ / 100µg
Description
MA1-10714 detects Neisseria gonorrhoeae. MA1-10714 has been successfully used in ELISA applications. The MA1-10714 immunogen is UV-inactivated Neisseria gonorrhoeae cells W-I, W-II, W-III.
Although pathogenic bacteria constitute only a small portion of bacterial species, infectious diseases caused by pathogenic bacteria affect millions of people worldwide. Diarrheal diseases, many of which are bacterial, are the second leading cause of death worldwide, claiming an estimated 5 million people annually. Furthermore, an estimated 3 million people die of tuberculosis annually, 500,000 people die of pertussis, and 25,000 die of typhoid. The ability of bacteria to utilize various food sources is a major factor behind their pervasiveness. Their vast numbers lead to rapid evolutionary changes resulting in emerging diseases as well as antibiotic-resistant strains of well known diseases. Examples of these are multi-drug resistant tuberculosis and methicillin-resistant Staphylococcus aureus. Research into the detection and treatment of these infectious disease agents will continue to expand in an effort to alleviate the effects of these diseases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neisseria gonorrhoeae
Monoclonal
0.4 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein G
RUO
Bacteria
Antibody
IgG3
ELISA
GC12-316.4
Unconjugated
Gonococci; N. gonorrhoeae
UV-inactivated Neisseria gonorrhoeae cells W-I, W-II, W-III.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles