Learn More
NEK11 Monoclonal Antibody (OTI5E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501608S
Description
NEK11 Monoclonal antibody specifically detects NEK11 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| NEK11 | |
| Monoclonal | |
| 0.76 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NCE2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 64419 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI5E9 | |
| Unconjugated | |
| MTMR14 | |
| 4932416N14Rik; Nek11; never in mitosis A-related kinase 11; NIMA (never in mitosis gene a)- related kinase 11; NIMA (never in mitosis gene a)-related expressed kinase 11; NIMA related kinase 11; NIMA-related kinase 11; nimA-related protein kinase 11; Serine/threonine-protein kinase Nek11 | |
| Full length human recombinant protein of human MTMR14 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.