Learn More
Weigert™ Neodisher™ S Defoamer
Product Code. 10202151
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10202151 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10202151 Supplier Weigert™ Supplier No. 430148
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
To prevent foaming in alkaline cleaning solutions which are highly polluted by fat, protein, starch, sugar and others. Weigert™ Neodisher™ S Defoamer can be used in machine pre-cleaning as well as in main cleaning.
  • Based on emulsified silicone oil
  • For use in clinical and industrial laboratories, and the food and cosmetics industries
  • Avoid overdosing to prevent adhesion of silicone residues to surfaces
Specifications
Product Type Neodisher Antimousse
Volume (Metric) 1 L