missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) oxide, REacton™, 99.997% (REO), Thermo Scientific™

139.00€ - 497.00€

Chemical Identifiers

CAS 1313-97-9
MDL Number MFCD00011134
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15491047
View Documents
Thermo Scientific Alfa Aesar
013837.14
25g
139.00€
25g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15401057
View Documents
Thermo Scientific Alfa Aesar
013837.22
100g
497.00€
100g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1313-97-9
MFCD00011134

Specifications

1313-97-9
14,6450
336.48
Reacton™
(REO)
MFCD00011134
Insoluble in water. Soluble in acids.
99.997%
7.24 g/mL
Neodymium(III) oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-214-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.