missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon,Nepsilon,Nepsilon-Trimethyl-d9-L-lysine HCl, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N6,N6,N6-Trimethyl-L-lysine-D9 (N,N,N-trimethyl-D9) hydrochloride |
| Molecular Formula | C9 D9 H11 N2 O2 |
| Quantity | 0.01 g |
| Synonym | 1-Pentanaminium, 5-amino-5-carboxy-N,N,N-tri(methyl-d3)-, inner salt, (5 S)- (ACI), N6,N6,N6-Trimethyl-L-lysine-D9 (N,N,N-trimethyl-D9), Lysine betaine-d9, Nε,Nε,Nε-Trimethyllysine-d9, N6,N6,N6-Tri(methyl-d3)-L-lysine inner salt, (5 S)-5-Amino-5-carboxy-N,N,N-tri(methyl-d3)-1-pentanaminium inner salt, (5 S)-5-Amino-5-carboxy-N,N,N-tri(methyl-d3)-1-pentanaminium inner salt, (5 S)-5-Amino-5-carboxy-N,N,N-tri(methyl-d3)-1-pentanaminium inner salt |
| InChI Formula | InChI=1 S/C9H20N2O2/c1-11(2,3)7-5-4-6-8(10)9(12)13/h8H,4-7,10H2,1-3H3/t8-/m0/s1/i1D3,2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])[N+](CCCC[C@H](N)C(=O)[O-])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | (2 S)-2-amino-6-[tris(trideuteriomethyl)azaniumyl]hexanoate |
| Molecular Weight (g/mol) | 197.323 |
| Formula Weight | 197.209 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction