Learn More
NET1 Antibody (CL3063) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46648MFV450
Product Code. 30299198
  • 759.00€ / 0.10mL
Description
NET1 Monoclonal antibody specifically detects NET1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NET1
Monoclonal
mFluor Violet 450 SE
ARHGEF8neuroepithelial cell-transforming gene 1 protein, guanine nucleotide regulatory protein (oncogene), NET1A, neuroepithelial cell transforming 1, neuroepithelial cell transforming gene 1, neuroepithelioma transforming gene 1, p65 Net1 proto-oncogene protein, Proto-oncogene p65 Net1, Rho guanine nucleotide exchange factor (GEF) 8, Rho guanine nucleotide exchange factor 8, small GTP-binding protein regulator
This antibody was developed using a recombinant protein derived from P23975, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Mouse, Rat
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL3063
50mM Sodium Borate
Mouse
Protein A purified
RUO
10276
Store at 4⪚C in the dark.