Learn More
Rogo Sampaic™ Neubaeur Cell
Product Code. 11740845
Change view
Click to view available options
Depth (Metric):
0.1mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
11740845 0.1mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11740845 Supplier Rogo Sampaic™ Supplier No. ZMB050
  • ONLINE EXCLUSIVE PRICE
    123.00€
    115.70€ / Pack of 1
    Save 7.30€
    5% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Neubaeur Cell
Grid Type Double
Quantity 1Each
Sample Type Blood
Depth (Metric) 0.1mm