Learn More
NeuN Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26975-1-AP-20UL
 View more versions of this product
Product Code. 16865295
  • 145.00€ / 20µL
Description
NeuN (Fox-3, NEUN, HRNBP3, RBFOX3, RNA binding protein, fox-1) or Fox-1 homolog C, is a 350 amino acid protein that contains one RRM (RNA recognition motif) domain. Localized to both the nucleus and cytoplasm, Fox-3 is suggested to regulate alternative splicing events. Fox-3 is encoded by a gene located on human chromosome 17, which is comprised over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintece of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with maligt cell growth and LiFraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determit of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
Specifications
Specifications
NeuN
Polyclonal
Unconjugated
RBFOX3
FOX3, Homo sapiens (Human), HRNBP3, NeuN, RBFOX3
Rabbit
Antigen Affinity Chromatography
RUO
110256156, 146713, 287847, 52897
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A6NFN3, Q8BIF2
Rbfox3
NeuN Fusion Protein Ag25689
20 μL
Primary
Pig, Human, Mouse, Rat