missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies NeuroCult™ SM1 Without Insulin

Codice prodotto. 17138291
Encompass
Cambia vista
Click to view available options
Quantity:
10 mL
Dimensione della confezione:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
17138291 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17138291 Fornitore STEMCELL Technologies N. del fornitore 05733

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Serum-free neural supplement without insulin

  • NeuroCult™ SM1 Without Insulin is an optimized serum-free culture supplement, based on the published B27 formulation (Brewer et al.), with insulin removed
  • NeuroCult™ SM1 Without Insulin can be used in protocols where the effects of insulin are being studied, or where insulin may interfere with cellular function
  • In combination with a basal medium of choice, NeuroCult™ SM1 Without Insulin can be used for neural progenitor cell expansion, the study of neural stem and progenitor cell differentiation, and cardiomyocyte and hepatocyte differentiation protocols

Specifica

Cell Type Neural Stem and Progenitor Cells, Neurons, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation, Expansion, Maintenance
Quantity 10 mL
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.