missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofibromin 1 Antibody (CL11865) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-15123DL594
This item is not returnable.
View return policy
Description
Neurofibromin 1 Monoclonal antibody specifically detects Neurofibromin 1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Neurofibromin 1 | |
| Monoclonal | |
| DyLight 594 | |
| DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS | |
| This antibody was developed using a synthetic peptide derived from P21359, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| 4763 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunocytochemistry/Immunofluorescence | |
| CL11865 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction