missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurolysin Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18401FR
This item is not returnable.
View return policy
Description
Neurolysin Polyclonal antibody specifically detects Neurolysin in Human samples. It is validated for Western Blot
Specifications
| Neurolysin | |
| Polyclonal | |
| Western Blot | |
| AGTBPFLJ23002, angiotensin binding protein, Angiotensin-binding protein, DKFZp564F123, EC 3.4.24, EC 3.4.24.16, KIAA1226EP24.16, MEP, mitochondrial, MOP, neurolysin (metallopeptidase M3 family) | |
| Recognizes region between residues Between Between 25 and 75. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 57486 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction