missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuronatin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26905-1-AP-20UL
This item is not returnable.
View return policy
Description
NNAT may participate in the maintece of segment identity in the hindbrain and pituitary development, and maturation or maintece of the overall structure of the nervous system. may function as a regulatory subunit of ion channels.Specifications
| Neuronatin | |
| Polyclonal | |
| Unconjugated | |
| NNAT | |
| neuronatin, NNAT, Peg5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4826 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16517 | |
| NNAT | |
| Neuronatin Fusion Protein Ag25408 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction