missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropilin-2 Antibody (96009R) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB567RI
This item is not returnable.
View return policy
Description
Neuropilin-2 Monoclonal antibody specifically detects Neuropilin-2 in Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF
Specifications
| Neuropilin-2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry, CyTOF | |
| 96009R | |
| Western Blot, Simple Western, Flow Cytometry, CyTOF-ready | |
| MGC126574, neuropilin 2, neuropilin-2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17) | |
| Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-2, Gln23-Asp857 (Val809-Asp825 del), Accession # O35276 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Hypoxia | |
| 8828 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction