Learn More
NeutraKine™ IL-3 Mouse anti-Human, Unconjugated, Clone: 3C11G7, Proteintech
Mouse Monoclonal Antibody
149.00€ - 319.00€
Specifications
| Antigen | NeutraKine™ IL-3 |
|---|---|
| Clone | 3C11G7 |
| Applications | Neutralization |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16820416
|
Proteintech
69004-1-IG-25UG |
25 μg |
149.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16810416
|
Proteintech
69004-1-IG-100UG |
100 μg |
319.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NeutraKine™ IL-3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-3. This antibody recognizes human antigen. The NeutraKine™ IL-3 antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
| NeutraKine™ IL-3 | |
| Neutralization | |
| Unconjugated | |
| Human | |
| IgG1 |
| 3C11G7 | |
| Monoclonal | |
| Mouse | |
| 3562 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.