missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutrophil Elastase/ELA2 Antibody (950317R) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB91671RF
This item is not returnable.
View return policy
Description
Neutrophil Elastase/ELA2 Monoclonal antibody specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| Neutrophil Elastase/ELA2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 950317R | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Neutrophil Elastase/ELA2, Met1-Asn252, Accession # P08246 | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 1991 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction