missing translation for 'onlineSavingsMsg'
Learn More

NF 279, TRC

Product Code. 30004620
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004620 10 mg 10mg
30011870 5 mg 5mg
30003819 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004620 Supplier Toronto Research Chemicals Supplier No. TRCN39168010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202983-32-2
Molecular Formula C49 H30 N6 O23 S6 . 6Na
Molecular Weight (g/mol) 1401.12
Synonym NF 279, 8,8'-[Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)]bis-1,3,5-naphthalenetrisulfonic Acid Hexasodium Salt
IUPAC Name hexasodium;8-[[4-[[4-[[4-[[4-[(4,6,8-trisulfonato-1-naphthyl)carbamoyl]phenyl]carbamoyl]phenyl]carbamoylamino]benzoyl]amino]benzoyl]amino]naphthalene-1,3,5-trisulfonate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1cc(c2c(NC(=O)c3ccc(NC(=O)c4ccc(NC(=O)Nc5ccc(cc5)C(=O)Nc6ccc(cc6)C(=O)Nc7ccc(c8cc(cc(c78)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])cc4)cc3)ccc(c2c1)S(=O)(=O)[O-])S(=O)(=O)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C49H36N6O23S6.6Na/c56-45(50-29-9-1-27(2-10-29)47(58)54-37-17-19-39(81(67,68)69)35-21-33(79(61,62)63)23-41(43(35)37)83(73,74)75)25-5-13-31(14-6-25)52-49(60)53-32-15-7-26(8-16-32)46(57)51-30-11-3-28(4-12-30)48(59)55-38-18-20-40(82(70,71)72)36-22-34(80(64,65)66)24-42(44(36)38)84(76,77)78;;;;;;/h1-24H,(H,50,56)(H,51,57)(H,54,58)(H,55,59)(H2,52,53,60)(H,61,62,63)(H,64,65,66)(H,67,68,69)(H,70,71,72)(H,73,74,75)(H,76,77,78);;;;;;/q;6*+1/p-6
Formula Weight 1399.9073
Chemical Name or Material 8,8'-[Carbonylbis(imino-4,1-phenylenecarbonylimino-4,1-phenylenecarbonylimino)]bis-1,3,5-naphthalenetrisulfonic acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.