missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NF 449

Description
NF 449 is a potent purinergic receptor antagonist that displays high selectivity for P2X1 (IC50 values are 0.28, 0.69, 120, 1820, 47000 and > 300000 nM for rP2X1, rP2X1+5, rP2X2+3, rP2X3, rP2X2 and P2X4 receptors respectively). Provides antithrombotic protection in vivo. Also acts as a Gsα-selective antagonist. Also inhibits DNA-binding activity of HMGA2 (IC50 = 0.43 μM).
This product is supplied with a high degree of hydration and some residual NaCl, the amount of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content and the maximum solubility threshold to use in dilution calculations.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 1505.06 |
| Purity | 90% |
| Molecular Weight (g/mol) | 1439.109 |
| Chemical Name or Material | NF 449 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?