Learn More
Invitrogen™ NF2 Recombinant Rabbit Monoclonal Antibody (JE31-02)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550810
Description
Positive control: Hela cell lysate, mouse brain tissue lysate, rat brain tissue lysate. Predicted band size: 70 kDa Subcellular Location: Cell membrane, Cell projection, Cytoplasm, Cytoskeleton, Membrane, Nucleus.
NF2 encodes a protein, Merlin, that is similar to some members of the ERM family of proteins that are thought to link cytoskeletal components with proteins in the cell membrane. Merlin has been shown to interact with cell-surface proteins, proteins involved in cytoskeletal dynamics and proteins involved in regulating ion transport. This gene is expressed at high levels during embryonic development; in adults, significant expression is found in Schwann cells, meningeal cells, lens and nerve. Mutations in this gene are associated with neurofibromatosis type II which is characterized by nervous system and skin tumors and ocular abnormalities. Two predominant isoforms and a number of minor isoforms are produced by alternatively spliced transcripts.
Specifications
| NF2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P35240, P46662, Q63648 | |
| Nf2 | |
| Synthetic peptide within Human Merlin aa 546-595/595. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| JE31-02 | |
| Unconjugated | |
| Nf2 | |
| ACN; BANF; Merlin; MGC123124; moesin-ezrin-radixin like; moesin-ezrin-radixin-like protein; moesin-ezrin-radizin-like protein; neurofibromatosis 2; neurofibromin 2; neurofibromin 2 (bilateral acoustic neuroma); neurofibromin 2 (merlin); neurofibromin 2b (merlin); neurofibromin-2; Nf2; Nf-2; NF2a; nf2b; OTTHUMP00000199607; SCH; Schwannomerlin; schwannomin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18016, 25744, 4771 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.