missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF45 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | NF45 |
|---|---|
| Clone | 1A3A2 |
| Concentration | 1200 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235488
|
Proteintech
67685-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235355
|
Proteintech
67685-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NF45 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NF45. This antibody recognizes human, mouse, rat antigen. The NF45 antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.Specifications
| NF45 | |
| 1200 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3608 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1A3A2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| MGC8391; NF45; PRO3063 | |
| IgG2b | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title