missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFATC2IP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00084901-D01P
This item is not returnable.
View return policy
Description
NFATC2IP Polyclonal antibody specifically detects NFATC2IP in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| NFATC2IP | |
| Polyclonal | |
| Western Blot 1:500, Immunocytochemistry/ Immunofluorescence | |
| AAH21551.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 84901 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.4) | |
| 45 kDa NF-AT-interacting protein, ESC2, FLJ14639, MGC126790, MGC138387, NFATC2-interacting protein, NIP4545 kDa NFAT-interacting protein, Nuclear factor of activated T-cells, cytoplasmic 2-interacting protein, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2interacting protein, RAD60 | |
| NFATC2IP (AAH21551.1, 1 a.a. - 138 a.a.) full-length human protein. MSEPLQSVVDHMATHLGVSPSRILLLFGETELSPTATPRTLKLGVADIIDCVVLTSSPEATETSQQLQLRVQGKEKHQTLEVSLSRDSPLKTLMSHYEEAMGLSGRKLSFFFDGTKLSGRELPADLGMESGDLIEVWG | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction