missing translation for 'onlineSavingsMsg'
Learn More

NFkB p105/p50, p Ser337 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

258.00€ - 398.00€

Specifications

Antigen NFkB p105/p50 (p Ser337)
Concentration 1.0 mg/mL
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18281903
View Documents
Novus Biologicals
NB100-82074-0.025ML
0.025 mL
258.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18234700
View Documents
Novus Biologicals
NB100-82074
0.05 mL
398.00€
0.05mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NFkB p105/p50 Polyclonal specifically detects NFkB p105/p50 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

NFkB p105/p50 (p Ser337)
Polyclonal
Rabbit
Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
P19838
4790
The antiserum was produced against synthesized phosphopeptide derived from human NF-kB p105/p50 around the phosphorylation site of serine 337 (R-K-SP-D-L).
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
1.0 mg/mL
Unconjugated
RUO
Human, Rat, Mouse
DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50
NFKB1
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.