missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-13 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Brand: R&D Systems AF-413-SP
This item is not returnable.
View return policy
Description
IL-13 Polyclonal specifically detects IL-13 in Mouse samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| IL-13 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3596 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.05-0.15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P20109 | |
| IL13 | |
| E. coli-derived recombinant mouse IL-13 Ser26-Phe131 Accession # P20109 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-13 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction