missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ NHE3/SLC9A3 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 6550 |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP3-00589. |
| Gene Alias | isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 |
| Gene Symbol | SLC9A3 |
| Quantity | 0.1 mg |
| Research Category | Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?