Learn More
NHEJ1 Monoclonal Antibody (OTI2C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502006
Encompass_Preferred
 View more versions of this product
Product Code. 17505741
  • 540.87€ / 100µL
Description
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.
Specifications
Specifications
NHEJ1
Monoclonal
0.55 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9H9Q4
NHEJ1
Full length human recombit protein of human NHEJ1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2C8
Unconjugated
NHEJ1
XLF
Mouse
Affinity Chromatography
RUO
79840
-20° C, Avoid Freeze/Thaw Cycles