missing translation for 'onlineSavingsMsg'
Learn More

Nickel, Ni/Cr/Mo/N, AL-6XN, N08367, ARMI∣MBH

Product Code. 30326163
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326163 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326163 Supplier ARMI/MBH Supplier No. MBH13X NSA11 AC
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ARMI/MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. Their offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium∣Carbon∣Chromium∣Cobalt∣Copper∣Grade∣Manganese∣Molybdenum∣Nickel∣Niobium∣Nitrogen∣Phosphorus∣Silicon∣Tungsten∣Vanadium
Quantity 50 g
Weight 50 g
Form Pieces
Concentration or Composition (by Analyte or Components) Carbon 0.0159 %∣Phosphorus 0.0186 %∣Aluminium 0.02 %∣Tungsten 0.038 %∣Vanadium 0.0513 %∣Cobalt 0.0981 %∣Niobium 0.15 %∣Copper 0.187 %∣Nitrogen 0.203 %∣Silicon 0.275 %∣Manganese 0.64 %∣Molybdenum 6.16 %∣Chromium 20.19 %∣Nickel 23.89 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.