missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Mo/Cr, Alloy C276, N10276, 100g, ARMI∣MBH
Description
Nickel, Ni/Mo/Cr, Alloy C276, N10276, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Carbon/Titanium/Phosphorus/Magnesium/Silicon/Vanadium/Niobium/Aluminium/Copper/Cobalt/Manganese/Tungsten/Iron/Molybdenum/Chromium/Nickel |
| CAS | 7440-02-0 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.002 %/Carbon 0.0032 %/Titanium 0.007 %/Phosphorus 0.008 %/Magnesium 0.01 %/Silicon 0.017 %/Vanadium 0.018 %/Niobium 0.061 %/Aluminium 0.114 %/Copper 0.14 %/Cobalt 0.17 %/Manganese 0.42 %/Tungsten 3.23 %/Iron 6.09 %/Molybdenum 15.4 %/Chromium 15.9 %/Nickel 58.5 % |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?