Learn More
Nickel Electro™ Nickel Dual Spoon/Spatula Combo Utensils
Product Code. 11716567 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
120 mm
150 mm
180 mm
210 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11716567 120 mm Each
11726567 150 mm Each
11736567 180 mm Each
11746567 210 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11716567
  • ONLINE EXCLUSIVE PRICE
    14.15€
    12.21€ / Pack of 1
    Save 1.94€
    13% Off
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Use one tool for two jobs. Nickel Electro™ Nickel Dual Spoon/Spatula Combo Utensils measure liquids and powders with the spoon end and divide and separate materials with the spatula end.
Dual-use utensil measures liquids and fine powders with the spoon end; divides and separates materials with the spatula end.
  • Manufactured from Nickel.
Specifications
Quantity 1
Capacity (Metric) 2 mL
Product Type Spatula
Width (Metric) 20 mm
Dimensions (L x W) 120 x 20 mm
Material Nickel
Length (Metric) 120 mm