Learn More
Bochem™ Nickel Plated Brass Pinch Cock
Used to control the flow of liquid through flexible tubing
Brand:  Bochem™ 12535
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10166320
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • With flap
  • Manufactured from Nickel Plated Brass
Specifications
Specifications
Hoffman Pinchcock
Nickel Plated Brass
40 mm