missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Nickel Plated Brass Pinch Cock
Used to control the flow of liquid through flexible tubing
5.70€ - 85.85€
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11815101
|
Bochem™
12534 |
30 mm |
76.00€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11825101
|
Bochem™
12535 |
40 mm |
85.85€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10425241
|
Bochem™
12540 |
50 mm |
14.45€
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
10166320
|
Bochem™
12535 |
40 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- With flap
- Manufactured from Nickel Plated Brass
Specifications
Hoffman Pinchcock | |
Nickel Plated Brass |