missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide N-oxide, 98%

Product Code. 15457808
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457808 25 g 25g
15467808 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457808 Supplier Thermo Scientific Alfa Aesar Supplier No. H61292.14
  • ONLINE EXCLUSIVE PRICE
    238.00€
    214.65€ / 25g
    Save 23.35€
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1986-81-8 | C6H6N2O2 | 138.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1986-81-8
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00006202
InChI Key USSFUVKEHXDAPM-UHFFFAOYSA-N
PubChem CID 72661
IUPAC Name 1-oxidopyridin-1-ium-3-carboxamide
SMILES C1=CC(=C[N+](=C1)[O-])C(=O)N

Specifications

Melting Point 291°C to 293°C (decomposition)
Quantity 25 g
Sensitivity Hygroscopic
Synonym nicotinamide n-oxide, nicotinamide 1-oxide, 1-oxynicotinamide, 3-pyridinecarboxamide, 1-oxide, 1-oxy-nicotinamide, niacinamide-n-oxide, nicotinamide, 1-oxide, 3-pyridinecarboxamide,1-oxide, 3-carbamoylpyridin-1-ium-1-olate, 3-aminocarbonyl pyridinium-1-olate
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material Nicotinamide N-oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.