missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Nicotine-d4 (pyridine-d4), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Nicotine-D4 |
| CAS | 350818-69-8 |
| Molecular Formula | C10 2H4 H10 N2 |
| Quantity | 0.05 g |
| Synonym | 5-(1-Methyl-2-pyrrolidinyl)pyridine-2,3,4,6-d4, rac-Nicotine-D4, racemic Nicotine-D4 |
| InChI Formula | InChI=1 S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10 H,3,5,7H2,1H3/i2D,4 D,6 D,8 D |
| SMILES | [2 H]c1nc([2 H])c(C2CCCN2C)c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(1-methylpyrrolidin-2-yl)pyridine |
| Molecular Weight (g/mol) | 166.26 |
| Formula Weight | 166.1408 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction