missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine Receptor beta 4 Antibody (7F5B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 562.00€
Specifications
| Antigen | Nicotinic Acetylcholine Receptor beta 4 |
|---|---|
| Clone | 7F5B2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18234721
|
Novus Biologicals
NBP2-61742 |
0.1 mL |
562.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644609
|
Novus Biologicals
NBP2-61742-0.025ml |
0.025 mL |
203.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicotinic Acetylcholine Receptor beta 4 Monoclonal specifically detects Nicotinic Acetylcholine Receptor beta 4 in Human samples. It is validated for Western Blot, ELISA.Specifications
| Nicotinic Acetylcholine Receptor beta 4 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| cholinergic receptor, nicotinic, beta 4, cholinergic receptor, nicotinic, beta polypeptide 4, neuronal acetylcholine receptor subunit beta-4, neuronal nicotinic receptor beta 4 subunit | |
| CHRNB4 | |
| IgG1 | |
| Protein G purified |
| 7F5B2 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Cellular Markers, Neuronal Cell Markers | |
| 1143 | |
| Purified recombinant fragment of human CHRNB4 (AA: extra 22-236) expressed in E. Coli. | |
| Primary | |
| 56.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title