missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinuric-d4 Acid (pyridyl-d4), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Nicotinuric Acid-d4 |
| CAS | 1216737-36-8 |
| Molecular Formula | C8 2H4 H4 N2 O3 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C8H8N2O3/c11-7(12)5-10-8(13)6-2-1-3-9-4-6/h1-4 H,5H2,(H,10,13)(H,11,12)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1nc([2 H])c(C(=O)NCC(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | 2-[(2,4,5,6-tetradeuteriopyridine-3-carbonyl)amino]acetic acid |
| Molecular Weight (g/mol) | 184.19 |
| Formula Weight | 184.0786 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction