missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nidogen-1/Entactin Antibody (ELM1), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-08237UV
This item is not returnable.
View return policy
Description
Nidogen-1/Entactin Monoclonal antibody specifically detects Nidogen-1/Entactin in Mouse, Guinea Pig samples. It is validated for ELISA
Specifications
| Nidogen-1/Entactin | |
| Monoclonal | |
| DyLight 350 | |
| enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1 | |
| Murine EHS laminin preparation | |
| 100 μg | |
| Primary | |
| Mouse, Guinea Pig | |
| Purified |
| ELISA | |
| ELM1 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 4811 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction