missing translation for 'onlineSavingsMsg'
Learn More

Niflumic acid, 99+%, Thermo Scientific™

50.40€ - 288.00€

Chemical Identifiers

CAS 4394-00-7
Molecular Formula C13H9F3N2O2
Molecular Weight (g/mol) 282.22
MDL Number MFCD00010569
InChI Key JZFPYUNJRRFVQU-UHFFFAOYSA-N
Synonym niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french
PubChem CID 4488
ChEBI CHEBI:34888
IUPAC Name 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15483259
SDS View Documents
Thermo Scientific Alfa Aesar
J60489.06
5g
ONLINE EXCLUSIVE PRICE
51.40€
50.40€ / 5g
Save 1.00€
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15493259
SDS View Documents
Thermo Scientific Alfa Aesar
J60489.09
10g
ONLINE EXCLUSIVE PRICE
86.50€
84.79€ / 10g
Save 1.71€
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15403269
SDS View Documents
Thermo Scientific Alfa Aesar
J60489.18
50g
ONLINE EXCLUSIVE PRICE
294.00€
288.00€ / 50g
Save 6.00€
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a selective Cox-2 inhibitor and GPR35 activator. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.

Solubility
Soluble in ethanol (∼50 mg/ml), acetone (50 mg/ml, Clear to Slightly Hazy, Yellow), methanol (∼50 mg/ml), DMSO (56 mg/ml at 25°C), and acetonitrile (∼50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Hygroscopic. Keep away fom strong oxidizing agents.
Specifications

Chemical Identifiers

4394-00-7
282.22
JZFPYUNJRRFVQU-UHFFFAOYSA-N
4488
2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
C13H9F3N2O2
MFCD00010569
niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french
CHEBI:34888
OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F

Specifications

4394-00-7
MFCD00010569
Light sensitive
niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french
JZFPYUNJRRFVQU-UHFFFAOYSA-N
2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
4488
282.22
Niflumic acid
C13H9F3N2O2
UN2811
14,6531
Soluble in ethanol (∽50mg/ml),acetone (50mg/ml,Clear to Slightly Hazy,Yellow),methanol (∽50mg/ml),DMSO (56mg/ml at 25°C),and acetonitrile (∽50mg/ml).
OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F
282.22
CHEBI:34888
≥99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 224-516-2

missing translation for 'rtecsNumber' : QT2999100

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.