missing translation for 'onlineSavingsMsg'
Learn More

Nifurtimox, TRC

Product Code. 30136675
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136675 50 mg 50mg
30134944 25 mg 25mg
30011306 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136675 Supplier Toronto Research Chemicals Supplier No. TRCN45880050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23256-30-6
Molecular Formula C10 H13 N3 O5 S
Molecular Weight (g/mol) 287.29
Synonym 4-Thiomorpholinamine, 3-methyl-N-[(5-nitro-2-furanyl)methylene]-, 1,1-dioxide, Thiomorpholine, 3-methyl-4-[(5-nitrofurfurylidene)amino]-, 1,1-dioxide (7CI, 8CI), (±)-Nifurtimox, 3-Methyl-4-(5-nitrofurfurylideneamino)tetrahydro-1,4-thiazine-1,1-dioxide, 4-(5-Nitrofurfurylideneamino)-3-methylthiomorpholine 1,1-dioxide, BAY 2502, BAY-A 2502, Bayer 2502, Lampit, Nifurtimox, Tetrahydro-3-methyl-4-[(5-nitrofurfurylidene)amino]-4H-1,4-thiazine 1,1-dioxide
IUPAC Name (E)-N-(3-methyl-1,1-dioxo-1,4-thiazinan-4-yl)-1-(5-nitrofuran-2-yl)methanimine
SMILES CC1CS(=O)(=O)CCN1\N=C\c2oc(cc2)[N+](=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H13N3O5S/c1-8-7-19(16,17)5-4-12(8)11-6-9-2-3-10(18-9)13(14)15/h2-3,6,8H,4-5,7H2,1H3/b11-6+
Formula Weight 287.0576
Chemical Name or Material Nifurtimox
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.