missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nilotinib

Product Code. 15851032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15851032 250 mg 250mg
15841032 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15851032 Supplier Thermo Scientific Chemicals Supplier No. 462372500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 641571-10-0 | C28H22F3N7O | 529.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 641571-10-0
Molecular Formula C28H22F3N7O
Molecular Weight (g/mol) 529.53
InChI Key HHZIURLSWUIHRB-UHFFFAOYSA-N
IUPAC Name 4-methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}benzamide
SMILES CC1=CN(C=N1)C1=CC(NC(=O)C2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)=CC(=C1)C(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 529.52
Chemical Name or Material Nilotinib

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.