missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Nilotinib

Product Code. 17140202
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17140202 50 mg 50mg
17130202 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17140202 Supplier STEMCELL Technologies Supplier No. 73304
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Tyrosine kinase inhibitor; Inhibits BCR/ABL and ABL

  • Nilotinib is a second-generation inhibitor of the oncogenic tyrosine kinase BCR-ABL with IC50 values of 19, 140, and 9,200 nM for wild-type, E255K, and T315I mutant forms of BCR-ABL, respectively (Kitagawa et al.; Verstovsek et al.; O'Hare)
  • It binds to the ATP-binding pocket of ABL, with higher affinity than imatinib (Manley et al. 2006; Verstovsek et al.) It also has activity below 1 μM against discoidin domain receptors (DDR) -1 and -2, platelet-derived growth factor receptors (PDGFR) –α and -β, stem cell factor receptor (KIT), and colony-stimulating factor 1 receptor (CSF-1R; Manley et al. 2010)

Specifications

CAS 641571-10-0
Synonym AMN107; Tasigna
Quantity 50 mg
Molecular Formula C28H22F3N7O
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.