Learn More
Nilotinib Regio Isomer, TRC
Product Code. 30005482
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005482 5 mg 5mg
30004023 25 mg 25mg
30006474 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005482 Supplier Toronto Research Chemicals Supplier No. TRCN4653205MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 641571-15-5
Molecular Formula C28 H22 F3 N7 O
Molecular Weight (g/mol) 529.52
Synonym 4-methyl-N-[3-(5-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-, 4-Methyl-N-[3-(5-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]benzamide, 4-Methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-N-[5-(5-methyl-1H-imidazol-1-yl)-3-(trifluoromethyl)phenyl]benzamide, 4-Methyl-N-[3-(5-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzamide, Nilotinib isomer
IUPAC Name 4-methyl-N-[3-(5-methylimidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]benzamide
SMILES Cc1ccc(cc1Nc2nccc(n2)c3cccnc3)C(=O)Nc4cc(cc(c4)C(F)(F)F)n5cncc5C
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C28H22F3N7O/c1-17-5-6-19(10-25(17)37-27-34-9-7-24(36-27)20-4-3-8-32-15-20)26(39)35-22-11-21(28(29,30)31)12-23(13-22)38-16-33-14-18(38)2/h3-16H,1-2H3,(H,35,39)(H,34,36,37)
Formula Weight 529.1838
Chemical Name or Material Nilotinib regioisomer