missing translation for 'onlineSavingsMsg'
Learn More

Nilutamide, TRC

Product Code. 30135556
Change view
Click to view available options
Quantity:
10 mg
100 mg
2.5 g
Unit Size:
100mg
10mg
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135556 2.5 g 2.5g
30009418 10 mg 10mg
30008532 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135556 Supplier Toronto Research Chemicals Supplier No. TRCN4675002.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63612-50-0
Molecular Formula C12 H10 F3 N3 O4
Molecular Weight (g/mol) 317.22
Synonym Nilutamide, 5,5-Dimethyl-3-[4-nitro-3-(trifluoromethyl)phenyl]imidazolidine-2,4-dione
IUPAC Name 5,5-dimethyl-3-[4-nitro-3-(trifluoromethyl)phenyl]imidazolidine-2,4-dione
SMILES CC1(C)NC(=O)N(C1=O)c2ccc(c(c2)C(F)(F)F)[N+](=O)[O-]

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H10F3N3O4/c1-11(2)9(19)17(10(20)16-11)6-3-4-8(18(21)22)7(5-6)12(13,14)15/h3-5H,1-2H3,(H,16,20)
Formula Weight 317.0623
Chemical Name or Material Nilutamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.