missing translation for 'onlineSavingsMsg'
Learn More

Nimbin, TRC

Product Code. 30012817
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012817 25 mg 25mg
30011997 1 mg 1mg
30003343 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012817 Supplier Toronto Research Chemicals Supplier No. TRCN47628025MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5945-86-8
Molecular Formula C30 H36 O9
Molecular Weight (g/mol) 540.6
Synonym (4alpha,5alpha,6alpha,7alpha,15beta,17alpha)-6-(acetyloxy)-7,15:21,23-diepoxy-4,8-dimethyl-1-oxo-8,24-dinor-11,12-secochola-2,13,20,22-tetraene-4,11-dicarboxylic acid methyl ester, 18,24-Dinor-11,12-secochola-2,13,20,22-tetraene-4,11-dicarboxylic acid, 6-(acetyloxy)-7,15:21,23-diepoxy-4,8-dimethyl-1-oxo-, dimethyl ester, (4α,5α,6α,7α,15β,17α)-, 2H-Cyclopenta[b]naphtho[2,3-d]furan-10-acetic acid, 6α-carboxy-2α-(3-furyl)-3,3aα,4aβ,5,5aα,6,9,9a,10α,10a-decahydro-5α-hydroxy-1,6,9aβ,10aβ-tetramethyl-9-oxo-, dimethyl ester, acetate (8CI), 2H-Cyclopenta[b]naphtho[2,3-d]furan-10-acetic acid, 6β-carboxy-2α-(3-furyl)-3,3aβ,4aβ,5,5aα,6,9,9a,10,10a-decahydro-5α-hydroxy-1,6,9aβ,10aβ-tetramethyl-9-oxo-, dimethyl ester, acetate (7CI), Nimbin (6CI), 2H-Cyclopenta[b]naphtho[2,3-d]furan-10-acetic acid, 5-(acetyloxy)-2-(3-furanyl)-3,3a,4a,5,5a,6,9,9a,10,10a-decahydro-6-(methoxycarbonyl)-1,6,9a,10a-tetramethyl-9-oxo-, methyl ester, [2R-(2α,3aα,4aβ,5α,5aα,6α,9aβ,10β,10aβ)]-
SMILES COC(=O)C[C@@H]1[C@]2(C)[C@@H]([C@@H](OC(=O)C)[C@@H]3O[C@@H]4C[C@H](C(=C4[C@@]13C)C)c5cocc5)[C@](C)(OC(=O)C)C=CC2=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C30H36O9/c1-15-19(18-9-11-36-14-18)12-20-24(15)30(6)21(13-23(34)35-7)29(5)22(33)8-10-28(4,39-17(3)32)26(29)25(27(30)38-20)37-16(2)31/h8-11,14,19-21,25-27H,12-13H2,1-7H3/t19-,20-,21-,25-,26+,27+,28-,29+,30-/m1/s1
Formula Weight 540.2359
Percent Purity >95%
Chemical Name or Material Nimbin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.