missing translation for 'onlineSavingsMsg'
Learn More

Nipah Virus Glycoprotein G Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

201.00€ - 398.00€

Specifications

Antigen Nipah Virus Glycoprotein G
Dilution Western Blot 1:2000-1:20000, ELISA 1:5000-1:10000
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30281373
View Documents
Novus Biologicals
NBP3-43765-20ul
20 μL
201.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30281372
View Documents
Novus Biologicals
NBP3-43765-100ul
100 μL
398.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nipah Virus Glycoprotein G Polyclonal antibody specifically detects Nipah Virus Glycoprotein G in Virus samples. It is validated for ELISA,Western Blot
Specifications

Specifications

Nipah Virus Glycoprotein G
ELISA, Western Blot
Unconjugated
Rabbit
Virus
HeV-G
IgG
Antigen and protein A Affinity-purified
Western Blot 1:2000-1:20000, ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
0.2 μm filtered solution in PBS
Produced in rabbits immunized with purified, recombinant Nipah Virus Glycoprotein G (RefSeq#: AAY43916.1; Gln71-Thr602)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.