missing translation for 'onlineSavingsMsg'
Learn More

Niperoditine, TRC

Product Code. 30469621
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30469621 25 mg
30464349 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30469621 Supplier Toronto Research Chemicals Supplier No. TRCN47880025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84845-75-0
Molecular Formula C20H26N4O5S
Molecular Weight (g/mol) 434.51
Synonym N-(1,3-Benzodioxol-5-ylmethyl)-N'-[2-[[[5-[(dimethylamino)methyl]-2-furanyl]methyl]thio]ethyl]-2-nitro-1,1-ethenediamine
IUPAC Name 1-N'-(1,3-benzodioxol-5-ylmethyl)-1-N-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-2-nitroethene-1,1-diamine
SMILES CN(C)Cc1oc(CSCCNC(=C[N+](=O)[O-])NCc2ccc3OCOc3c2)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H26N4O5S/c1-23(2)11-16-4-5-17(29-16)13-30-8-7-21-20(12-24(25)26)22-10-15-3-6-18-19(9-15)28-14-27-18/h3-6,9,12,21-22H,7-8,10-11,13-14H2,1-2H3
Formula Weight 434.1624
Chemical Name or Material Niperoditine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.