missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIPSNAP3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10751-1-AP-20UL
This item is not returnable.
View return policy
Description
The NIPSNAP proteins comprise a family of evolutionarily well-conserved proteins with strong sequence similarity to the central portion of a protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. NIPSNAP2, a novel gene encoding a protein with tyrosine phosphorylation sites and a transmembrane domain, is co-amplified with EGFR in approximately 40% of glioblastomas, the most common and maligt form of central nervous system tumors. While NIPSNAP3B is highly expressed skeletal muscle, NIPSNAP3A mRNA levels are low. NIPSNAP3A protein is associated with plasma membrane and partially localized in rafts. NIPSNAP proteins have been suggested to be important in vesicular transport.Specifications
| NIPSNAP3A | |
| Polyclonal | |
| Unconjugated | |
| NIPSNAP3A | |
| HSPC299, NIPSNAP3A, NIPSNAP4, Protein NipSnap homolog 3A, Protein NipSnap homolog 4, TASSC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25934, 685080, 73398 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UFN0 | |
| Nipsnap3a | |
| NIPSNAP3A Fusion Protein Ag1143 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction